missing translation for 'onlineSavingsMsg'
Learn More

(2R,2'R,3R,3'R,4R)-2,2'-Bis[3,4-bis(phenylmethoxy)phenyl]-3,3',4,4'-tetrahydro-5,5',7,7'-tetrakis(phenylmethoxy)[4,8'-bi-2H-1-benzopyran]-3,3'-diyl Ester 3,4,5-Tris(phenylmethoxy)-benzoic Acid, TRC

Product Code. 30330274
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330274 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330274 Supplier Toronto Research Chemicals Supplier No. TRCB5226332.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R,2'R,3R,3'R,4R)-2,2'-Bis[3,4-bis(phenylmethoxy)phenyl]-3,3',4,4'-tetrahydro-5,5',7,7'-tetrakis(phenylmethoxy)[4,8'-bi-2H-1-benzopyran]-3,3'-diyl Ester 3,4,5-Tris(phenylmethoxy)-benzoic Acid
CAS 223387-33-5
Molecular Formula C142H118O20
Quantity 2.5 mg
Synonym Benzoic acid, 3,4,5-tris(phenylmethoxy)-, (2R,2′R,3R,3′R,4R)-2,2′-bis[3,4-bis(phenylmethoxy)phenyl]-3,3′,4,4′-tetrahydro-5,5′,7,7′-tetrakis(phenylmethoxy)[4,8′-bi-2H-1-benzopyran]-3,3′-diyl ester
InChI Formula InChI=1S/C142H118O20/c143-141(115-77-127(153-93-107-59-31-9-32-60-107)138(157-97-111-67-39-13-40-68-111)128(78-115)154-94-108-61-33-10-34-62-108)160-131-83-118-121(148-88-102-49-21-4-22-50-102)84-125(152-92-106-57-29-8-30-58-106)133(137(118)161-135(131)113-71-73-119(146-86-100-45-17-2-18-46-100)122(75-113)149-89-103-51-23-5-24-52-103)134-132-124(151-91-105-55-27-7-28-56-105)81-117(145-85-99-43-15-1-16-44-99)82-126(132)159-136(114-72-74-120(147-87-101-47-19-3-20-48-101)123(76-114)150-90-104-53-25-6-26-54-104)140(134)162-142(144)116-79-129(155-95-109-63-35-11-36-64-109)139(158-98-112-69-41-14-42-70-112)130(80-116)156-96-110-65-37-12-38-66-110/h1-82,84,131,134-136,140H,83,85-98H2/t131-,134-,135-,136-,140-/m1/s1
SMILES O(C(=O)C1=CC(OCC2=CC=CC=C2)=C(OCC3=CC=CC=C3)C(OCC4=CC=CC=C4)=C1)[C@@H]5[C@H](C=6C(O[C@@H]5C7=CC(OCC8=CC=CC=C8)=C(OCC9=CC=CC=C9)C=C7)=CC(OCC%10=CC=CC=C%10)=CC6OCC%11=CC=CC=C%11)C%12=C%13C(=C(OCC%14=CC=CC=C%14)C=C%12OCC%15=CC=CC=C%15)C[C@@H](OC(=O)C%16=CC(OCC%17=CC=CC=C%17)=C(OCC%18=CC=CC=C%18)C(OCC%19=CC=CC=C%19)=C%16)[C@H](O%13)C%20=CC(OCC%21=CC=CC=C%21)=C(OCC%22=CC=CC=C%22)C=C%20
IUPAC Name (2R,2'R,3R,3'R,4R)-5,5',7,7'-tetrakis(benzyloxy)-2,2'-bis(3,4-bis(benzyloxy)phenyl)-[4,8'-bichromane]-3,3'-diyl bis(3,4,5-tris(benzyloxy)benzoate)
Molecular Weight (g/mol) 2144.44
Formula Weight 2142.85
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.