missing translation for 'onlineSavingsMsg'
Learn More

Bendamustine Dimer Deschloro 2’-Allyl Ester, TRC

Product Code. 30331974
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331974 25 mg 25mg
30346229 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30331974 Supplier Toronto Research Chemicals Supplier No. TRCB13254025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bendamustine Dimer Deschloro 2’-Allyl Ester
Analyte or Component Names Bendamustine Dimer Deschloro 2’-Allyl Ester
Shipping Condition Room Temperature
Molecular Formula C28 H22 Cl2 F N O3
Synonym 4-(5-((2-((4-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoyl)oxy)ethyl)(2-hydroxyethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic Acid Allyl Ester
InChI Formula InChI=1S/C28H22Cl2FNO3/c1-28(2)21(15-22(30)17-8-11-19(29)12-9-17)26(28)27(33)35-25(16-32)18-10-13-23(31)24(14-18)34-20-6-4-3-5-7-20/h3-15,21,25-26H,1-2H3/b22-15-
Solution Type Neat
SMILES CC1(C)C(\C=C(/Cl)\c2ccc(Cl)cc2)C1C(=O)OC(C#N)c3ccc(F)c(Oc4ccccc4)c3
IUPAC Name [cyano-(4-fluoro-3-phenoxyphenyl)methyl] 3-[(Z)-2-chloro-2-(4-chlorophenyl)ethenyl]-2,2-dimethylcyclopropane-1-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 510.384
Quantity 25 mg
Formula Weight 509.096
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.