missing translation for 'onlineSavingsMsg'
Learn More

N1-(3-((1-Benzyl-1H-indazol-3-yl)oxy)propyl)-N1,N3,N3-trimethylpropane-1,3-diamine, TRC

Product Code. 30333785
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333785 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333785 Supplier Toronto Research Chemicals Supplier No. TRCB13149525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N1-(3-((1-Benzyl-1H-indazol-3-yl)oxy)propyl)-N1,N3,N3-trimethylpropane-1,3-diamine
CAS 1337966-15-0
Molecular Formula C23H32N4O
Quantity 25 mg
Synonym Benzydamine Hydrochloride Impurity D
InChI Formula InChI=1S/C23H32N4O/c1-25(2)15-9-16-26(3)17-10-18-28-23-21-13-7-8-14-22(21)27(24-23)19-20-11-5-4-6-12-20/h4-8,11-14H,9-10,15-19H2,1-3H3
SMILES CN(C)CCCN(C)CCCOc1nn(Cc2ccccc2)c3ccccc13
IUPAC Name N'-[3-(1-benzylindazol-3-yl)oxypropyl]-N,N,N'-trimethylpropane-1,3-diamine
Molecular Weight (g/mol) 380.53
Formula Weight 380.2576
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.