missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethyl(tetradec-5-yn-1-yloxy)silane, TRC

Product Code. 30337517
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337517 250 mg 250mg
30336470 100 mg 100mg
30342233 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30337517 Supplier Toronto Research Chemicals Supplier No. TRCB692525250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyldimethyl(tetradec-5-yn-1-yloxy)silane
Analyte or Component Names tert-Butyldimethyl(tetradec-5-yn-1-yloxy)silane
Shipping Condition Room Temperature
Molecular Formula C20 H40 O Si
InChI Formula InChI=1S/C20H40OSi/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-21-22(5,6)20(2,3)4/h7-13,16-19H2,1-6H3
Solution Type Neat
SMILES CCCCCCCCC#CCCCCO[Si](C)(C)C(C)(C)C
IUPAC Name tert-butyl-dimethyl-tetradec-5-ynoxysilane
Industry Type Pharmaceutical
Molecular Weight (g/mol) 324.616
Quantity 250 mg
Formula Weight 324.285
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.