missing translation for 'onlineSavingsMsg'
Learn More

1-Deoxygalactonojirimycin Hydrochloride, TRC

Product Code. 30350253
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350253 10 mg 10mg
30358381 50 mg 50mg
30357817 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350253 Supplier Toronto Research Chemicals Supplier No. TRCD23650010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75172-81-5
Molecular Formula C6 H13 N O4 . Cl H
Molecular Weight (g/mol) 199.63
Synonym 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride (1:1), (2R,3S,4R,5S)-, 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride, (2R,3S,4R,5S)- (9CI), 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride, [2R-(2α,3α,4α,5β)]-, 1-Deoxygalactonojirimycin hydrochloride, Migalastat hydrochloride
IUPAC Name (2R,3S,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES Cl.OC[C@H]1NC[C@H](O)[C@@H](O)[C@H]1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C6H13NO4.ClH/c8-2-3-5(10)6(11)4(9)1-7-3;/h3-11H,1-2H2;1H/t3-,4+,5+,6-;/m1./s1
Formula Weight 199.0611
Percent Purity >95%
Chemical Name or Material Deoxygalactonojirimycin hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.