missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Danofloxacin-d2 Formate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Danofloxacin-d2 Formate Salt |
| Molecular Formula | C19H18D2FN3O3 • x(HCOOH) |
| Quantity | 1 mg |
| Synonym | 1-Cyclopropyl-6-fluoro-1,4-dihydro-7-[(1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]hept-2-yl]-4-oxo-3-quinolinecarboxylic Acid-d2 Formate Salt, 2,5-Diazabicyclo[2.2.1]heptane 3-Quinolinecarboxylic Acid derivative-d2 Formate Salt, Advocin-d2 Formate Salt, Advocip-d2 Formate Salt |
| InChI Formula | InChI=1S/C19H20FN3O3.CH2O2/c1-21-7-12-4-11(21)8-22(12)17-6-16-13(5-15(17)20)18(24)14(19(25)26)9-23(16)10-2-3-10;2-1-3/h5-6,9-12H,2-4,7-8H2,1H3,(H,25,26);1H,(H,2,3)/t11-,12-;/m0./s1/i1D2; |
| SMILES | FC1=CC2=C(C=C1N3[C@@H]4CN(C([2H])[2H])[C@@H](C4)C3)N(C5CC5)C=C(C(O)=O)C2=O.[HCOOH] |
| IUPAC Name | 1-cyclopropyl-6-fluoro-7-((1S,4S)-5-(methyl-d2)-2,5-diazabicyclo[2.2.1]heptan-2-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid formic acid |
| Molecular Weight (g/mol) | 405.42 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?