missing translation for 'onlineSavingsMsg'
Learn More

(S)-Carbidopa Methyl Ester (>90%), TRC

Product Code. 30350529
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350529 50 mg 50mg
30358156 10 mg -
30366852 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350529 Supplier Toronto Research Chemicals Supplier No. TRCC17602550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52514-63-3
Molecular Formula C11 H16 N2 O4
Molecular Weight (g/mol) 240.256
Synonym (S)-α-Hydrazino-3,4-dihydroxy-α-methyl-benzenepropanoic acid methyl ester, (S)-alpha-Hydrazino-3,4-dihydroxy-alpha-methylbenzenepropanoic Acid Methyl Ester
IUPAC Name methyl (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoate
SMILES COC(=O)[C@](C)(Cc1ccc(O)c(O)c1)NN

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H16N2O4/c1-11(13-12,10(16)17-2)6-7-3-4-8(14)9(15)5-7/h3-5,13-15H,6,12H2,1-2H3/t11-/m0/s1
Formula Weight 240.111
Chemical Name or Material Methyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate (Carbidopa Methyl Ester)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.