missing translation for 'onlineSavingsMsg'
Learn More

Crotonoyl Chloride, TRC

Product Code. 30350867
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350867 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350867 Supplier Toronto Research Chemicals Supplier No. TRCC81879525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Crotonoyl Chloride
CAS 10487-71-5
Molecular Formula C4H5ClO
Quantity 25 g
Synonym Crotonic Acid Chloride, 2-Butenoyl Chloride, beta-Methylacryloyl Chloride
InChI Formula InChI=1S/C4H5ClO/c1-2-3-4(5)6/h2-3H,1H3
SMILES CC=CC(=O)Cl
IUPAC Name but-2-enoyl chloride
Molecular Weight (g/mol) 104.53
Formula Weight 104.0029
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.