missing translation for 'onlineSavingsMsg'
Learn More

4-((3-(4-(Cyclopropanecarbonyl)piperazine-1-carbonyl)-4-fluorophenyl)(hydroxy)methyl)phthalazin-1(2H)-one, TRC

Product Code. 30351146
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30351146 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351146 Supplier Toronto Research Chemicals Supplier No. TRCC99227825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1460272-53-0
Molecular Formula C24H23FN4O4
Quantity 25 mg
Synonym Olaparib Metabolite M15
InChI Formula InChI=1S/C24H23FN4O4/c25-19-8-7-15(21(30)20-16-3-1-2-4-17(16)22(31)27-26-20)13-18(19)24(33)29-11-9-28(10-12-29)23(32)14-5-6-14/h1-4,7-8,13-14,21,30H,5-6,9-12H2,(H,27,31)
SMILES OC(C1=NNC(=O)c2ccccc12)c3ccc(F)c(c3)C(=O)N4CCN(CC4)C(=O)C5CC5
IUPAC Name 4-[[3-[4-(cyclopropanecarbonyl)piperazine-1-carbonyl]-4-fluorophenyl]-hydroxymethyl]-2H-phthalazin-1-one
Molecular Weight (g/mol) 450.46
Chemical Name or Material 4-((3-(4-(Cyclopropanecarbonyl)piperazine-1-carbonyl)-4-fluorophenyl)(hydroxy)methyl)phthalazin-1(2H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.