missing translation for 'onlineSavingsMsg'
Learn More

4,4'-DDE, TRC

Product Code. 30351192
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351192 5 g 5g
30350986 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30351192 Supplier Toronto Research Chemicals Supplier No. TRCD1952055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-55-9
Molecular Formula C14 H8 Cl4
Molecular Weight (g/mol) 318.03
Synonym Benzene, 1,1'-(dichloroethenylidene)bis[4-chloro- (9CI), Ethylene, 1,1-dichloro-2,2-bis(p-chlorophenyl)- (7CI,8CI), 1,1'-(2,2-Dichloroethenylidene)bis[4-chlorobenzene], 1,1-Bis(4-chlorophenyl)-2,2-dichloroethene, 1,1-Bis(p-chlorophenyl)-2,2-dichloroethylene, 1,1-Dichloro-2,2-bis(p-chlorophenyl)ethylene, 1,1-Dichloro-2,2-di(p-chlorophenyl)ethylene, 1,1'-Dichloro-2,2-bis(4-chlorophenyl)ethane, 2,2-Bis(4-chlorophenyl)-1,1-dichloroethene, 2,2-Bis(4-chlorophenyl)-1,1-dichloroethylene, 2,2-Di(p-chlorophenyl)-1,1-dichloroethylene, 2,2-Dichloro-1,1-bis(4-chlorophenyl)ethylene, 4,4'-DDE, 4,4'-Dichlorodiphenyldichloroethylene, DDE, NSC 1153, p,p'-DDE, p,p'-Dichlorodiphenyldichloroethylene
IUPAC Name 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene
SMILES ClC(=C(c1ccc(Cl)cc1)c2ccc(Cl)cc2)Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H8Cl4/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8H
Formula Weight 315.938
Chemical Name or Material 4,4'-DDE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.