missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Desmethyl Toremifene Hydrochloride Salt, TRC
Specifications
Specifications
| Chemical Name or Material | N-Desmethyl Toremifene hydrochloride |
| CAS | 176671-73-1 |
| Molecular Formula | C25 H26 Cl N O . H Cl |
| Quantity | 25 mg |
| Synonym | 2-[4-[(1Z)-4-Chloro-1,2-diphenyl-1-buten-1-yl]phenoxy]-N-methylethanamine Hydrochloride Salt, (Z)-4-Chloro-1,2-diphenyl-1-[4-[2-(N-methylamino)ethoxy]phenyl]-1-butene Hydrochloride Salt, Fc 1200 Hydrochloride Salt, N-Demethyltoremifene Hydrochloride Salt |
| InChI Formula | InChI=1S/C25H26ClNO.ClH/c1-27-18-19-28-23-14-12-22(13-15-23)25(21-10-6-3-7-11-21)24(16-17-26)20-8-4-2-5-9-20;/h2-15,27H,16-19H2,1H3;1H/b25-24-; |
| SMILES | Cl.CNCCOc1ccc(cc1)\C(=C(\CCCl)/c2ccccc2)\c3ccccc3 |
| IUPAC Name | 2-[4-[(Z)-4-chloro-1,2-diphenyl-but-1-enyl]phenoxy]-N-methyl-ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 428.39 |
| Formula Weight | 427.147 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?