missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorobutoxy)tetrahydropyran, TRC

Product Code. 30351837
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351837 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351837 Supplier Toronto Research Chemicals Supplier No. TRCC36444525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(4-Chlorobutoxy)tetrahydropyran
CAS 41302-05-0
Molecular Formula C9H17ClO2
Quantity 25 g
Synonym 2-(4-Chlorobutoxy)tetrahydro-pyran, 1-Chloro-4-(tetrahydropyranyloxy)butane, 2-(4-Chlorobutoxy)tetrahydro-2H-pyran, 2-(4-Chlorobutyloxy)tetrahydropyran, 4-(Tetrahydro-2-pyranyloxy)-1-chlorobutane, 4-Chloro-1-(tetrahydropyranyloxy)butane, 4-Chloro-1-butanol Tetrahydropyranyl Ether, 4-Chlorobutanol Tetrahydropyranyl Ether, 4-Chlorobutyl Tetrahydropyranyl Ether
InChI Formula InChI=1S/C9H17ClO2/c10-6-2-4-8-12-9-5-1-3-7-11-9/h9H,1-8H2
SMILES ClCCCCOC1CCCCO1
IUPAC Name 2-(4-chlorobutoxy)oxane
Molecular Weight (g/mol) 192.68
Formula Weight 192.0917
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.