missing translation for 'onlineSavingsMsg'
Learn More

Dapagliflozin Methyl Acetate, TRC

Product Code. 30351945
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351945 100 mg 100mg
30354773 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30351945 Supplier Toronto Research Chemicals Supplier No. TRCD183610100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dapagliflozin Methyl Acetate
CAS 1632287-34-3
Molecular Formula C23 H27 Cl O7
Quantity 100 mg
Synonym ((2R,3S,4R,5R,6S)-6-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)methyl Acetate, Dapagliflozin Impurity-S2D(Mono acetylated)
InChI Formula InChI=1S/C23H27ClO7/c1-3-29-17-7-4-14(5-8-17)10-16-11-15(6-9-18(16)24)23-22(28)21(27)20(26)19(31-23)12-30-13(2)25/h4-9,11,19-23,26-28H,3,10,12H2,1-2H3/t19-,20-,21+,22-,23+/m1/s1
SMILES CCOc1ccc(Cc2cc(ccc2Cl)[C@@H]3O[C@H](COC(=O)C)[C@@H](O)[C@H](O)[C@H]3O)cc1
IUPAC Name [(2R,3S,4R,5R,6S)-6-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-3,4,5-trihydroxyoxan-2-yl]methyl acetate
Molecular Weight (g/mol) 450.909
Formula Weight 450.145
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.