missing translation for 'onlineSavingsMsg'
Learn More

(S)-[2-[[2-(2-Chlorobenzoyl)-4-nitrophenyl]amino]-1-methyl-2-oxoethyl]-carbamic Acid Phenylmethyl Ester, TRC

Product Code. 30352891
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352891 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352891 Supplier Toronto Research Chemicals Supplier No. TRCC36454310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-(2-((2-(2-Chlorobenzoyl)-4-nitrophenyl)amino)-1-methyl-2-oxoethyl)-carbamic Acid Phenylmethyl Ester
CAS 58662-82-1
Molecular Formula C24H20ClN3O6
Quantity 10 mg
Synonym (S)-Benzyl (1-((2-(2-chlorobenzoyl)-4-nitrophenyl)amino)-1-oxopropan-2-yl)carbamate
InChI Formula InChI=1S/C24H20ClN3O6/c1-15(26-24(31)34-14-16-7-3-2-4-8-16)23(30)27-21-12-11-17(28(32)33)13-19(21)22(29)18-9-5-6-10-20(18)25/h2-13,15H,14H2,1H3,(H,26,31)(H,27,30)/t15-/m0/s1
SMILES C[C@H](NC(=O)OCc1ccccc1)C(=O)Nc2ccc(cc2C(=O)c3ccccc3Cl)[N+](=O)[O-]
IUPAC Name benzyl N-[(2S)-1-[2-(2-chlorobenzoyl)-4-nitroanilino]-1-oxopropan-2-yl]carbamate
Molecular Weight (g/mol) 481.89
Formula Weight 481.1041
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.