missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-1,2-Cyclohexanediaminedinitrate Platinum, TRC

Product Code. 30353062
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353062 2.5 mg 2.5mg
30349372 10 mg 10mg
30359800 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353062 Supplier Toronto Research Chemicals Supplier No. TRCC9892552.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6 H14 N4 O6 Pt
Molecular Weight (g/mol) 433.277
Synonym (1R,2R)-1,2-trans-Cyclohexanediamine)dinitrate platinum(II)
IUPAC Name [(1R,6R)-8-nitrooxy-7λ4,9λ4-diaza-8λ4-platinabicyclo[4.3.0]nonan-8-yl] nitrate
SMILES [O-][N+](=O)O[Pt]1(O[N+](=O)[O-])[NH2][C@@H]2CCCC[C@H]2[NH2]1

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C6H14N2.2NO3.Pt/c7-5-3-1-2-4-6(5)8;2*2-1(3)4;/h5-6H,1-4,7-8H2;;;/q;2*-1;+2/t5-,6-;;;/m1.../s1
Formula Weight 433.056
Chemical Name or Material (1R,2R)-1,2-Cyclohexanediaminedinitrate Platinum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.