missing translation for 'onlineSavingsMsg'
Learn More

N,N-Di-Boc-9-Methylbiotin Methyl Ester;, TRC

Product Code. 30353664
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353664 10 mg 10mg
30356832 5 mg 5mg
30359260 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353664 Supplier Toronto Research Chemicals Supplier No. TRCD41572510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Di-Boc-9-Methylbiotin Methyl Ester
Analyte or Component Names N,N-Di-Boc-9-Methylbiotin Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C22 H36 N2 O7 S
Synonym 1,3-Boc-9-Methyl-(+)-Biotin Methyl Ester, (3aS,4S,6aR)-Di-tert-butyl 4-(5-Methoxy-4-methyl-5-oxopentyl)-2-oxotetrahydro-1H-thieno[3,4-d]imidazole-1,3(2H)-dicarboxylate
InChI Formula InChI=1S/C22H36N2O7S/c1-13(17(25)29-8)10-9-11-15-16-14(12-32-15)23(19(27)30-21(2,3)4)18(26)24(16)20(28)31-22(5,6)7/h13-16H,9-12H2,1-8H3/t13?,14-,15-,16-/m0/s1
Solution Type Neat
SMILES COC(=O)C(C)CCC[C@@H]1SC[C@H]2[C@@H]1N(C(=O)OC(C)(C)C)C(=O)N2C(=O)OC(C)(C)C
IUPAC Name ditert-butyl (3aS,4S,6aR)-4-(5-methoxy-4-methyl-5-oxopentyl)-2-oxo-3a,4,6,6a-tetrahydrothieno[3,4-d]imidazole-1,3-dicarboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 472.595
Quantity 10 mg
Formula Weight 472.224
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.