missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl N-Nitroso Methadone, TRC

Product Code. 30353714
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353714 2.5 mg 2.50mg
30368677 25 mg 25mg
30365139 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353714 Supplier Toronto Research Chemicals Supplier No. TRCD2997602.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Desmethyl N-Nitroso Methadone
Analyte or Component Names N-Desmethyl N-Nitroso Methadone
Purity Grade Notes HPLC
Shipping Condition Room Temperature
Molecular Formula C20 H24 N2 O2
Synonym N-methyl-N-(5-oxo-4,4-diphenylheptan-2-yl)nitrous Amide
InChI Formula InChI=1S/C20H24N2O2/c1-4-19(23)20(15-16(2)22(3)21-24,17-11-7-5-8-12-17)18-13-9-6-10-14-18/h5-14,16H,4,15H2,1-3H3
Solution Type Neat
SMILES CC(N(N=O)C)CC(C1=CC=CC=C1)(C(CC)=O)C2=CC=CC=C2
Industry Type Pharmaceutical
Molecular Weight (g/mol) 324.42
Quantity 2.5 mg
Formula Weight 324.1838
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.