missing translation for 'onlineSavingsMsg'
Learn More

(3Beta,17Alpha)-4-Chloro-3-hydroxy-17-methyl-18-norandrosta-4,13-diene-17-(methyl acetate), TRC

Product Code. 30353729
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353729 25 mg 25mg
30368646 10 mg 10mg
30364000 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353729 Supplier Toronto Research Chemicals Supplier No. TRCC41180525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C22H31ClO3
Molecular Weight (g/mol) 378.93
IUPAC Name ((3S,8R,9S,10R,17R)-4-chloro-3-hydroxy-10,17-dimethyl-2,3,6,7,8,9,10,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl)methyl acetate
SMILES [H][C@H]12[C@@](C(CC[C@@]3(C)COC(C)=O)=C3CC2)([H])CCC4=C(Cl)[C@@H](O)CC[C@@]41C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H31ClO3/c1-13(24)26-12-21(2)10-8-15-14-4-5-18-20(23)19(25)9-11-22(18,3)17(14)7-6-16(15)21/h14,17,19,25H,4-12H2,1-3H3/t14-,17-,19-,21-,22+/m0/s1
Formula Weight 378.2
Chemical Name or Material (3beta,17alpha)-4-Chloro-3-hydroxy-17-methyl-18-norandrosta-4,13-diene-17-(methyl acetate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.