missing translation for 'onlineSavingsMsg'
Learn More

Cyanamide-15N2, TRC

Product Code. 30353987
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353987 10 mg 10mg
30351661 5 mg 5mg
30355429 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353987 Supplier Toronto Research Chemicals Supplier No. TRCC95250310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21420-37-1
Molecular Formula C H2 15N2
Molecular Weight (g/mol) 44.027
IUPAC Name (15N)azanylformonitrile
SMILES [15NH2]C#[15N]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/CH2N2/c2-1-3/h2H2/i2+1,3+1
Formula Weight 44.016
Chemical Name or Material Cyanamide-15N2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.