missing translation for 'onlineSavingsMsg'
Learn More

Deoxymannojirimycin Hydrochloride, TRC

Product Code. 30354448
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354448 25 mg 25mg
30362750 10 mg 10mg
30358532 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354448 Supplier Toronto Research Chemicals Supplier No. TRCD24000025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73465-43-7
Molecular Formula C6 H13 N O4 . Cl H
Molecular Weight (g/mol) 199.63
Synonym 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride (1:1), (2R,3R,4R,5R)-, 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride, (2R,3R,4R,5R)- (9CI), 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride, [2R-(2α,3β,4α,5α)]-, 1-Deoxymannojirimycin hydrochloride, manno-1-Deoxynojirimycin hydrochloride
IUPAC Name (2R,3R,4R,5R)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES Cl.OC[C@H]1NC[C@@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H13NO4.ClH/c8-2-3-5(10)6(11)4(9)1-7-3;/h3-11H,1-2H2;1H/t3-,4-,5-,6-;/m1./s1
Formula Weight 199.0611
Percent Purity >95%
Chemical Name or Material Deoxymannojirimycin, Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.