missing translation for 'onlineSavingsMsg'
Learn More

rac-trans-4-Defluoro-4-ethoxy Paroxetine Hydrochloride, TRC

Product Code. 30354627
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354627 1 mg 1mg
30358412 2.5 mg 2.5mg
30354628 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354627 Supplier Toronto Research Chemicals Supplier No. TRCD2289851MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346597-97-4
Molecular Formula C21 H25 N O4 . Cl H
Molecular Weight (g/mol) 391.89
Synonym (3R,4S)-rel-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-ethoxyphenyl)piperidine Hydrochloride (1:1), SB-242673A, Ph Eur Paroxetine Hydrochloride Impurity C
IUPAC Name (3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-ethoxyphenyl)piperidine;hydrochloride
SMILES Cl.CCOc1ccc(cc1)[C@@H]2CCNC[C@H]2COc3ccc4OCOc4c3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H25NO4.ClH/c1-2-23-17-5-3-15(4-6-17)19-9-10-22-12-16(19)13-24-18-7-8-20-21(11-18)26-14-25-20;/h3-8,11,16,19,22H,2,9-10,12-14H2,1H3;1H/t16-,19-;/m0./s1
Formula Weight 391.155
Percent Purity >95%
Chemical Name or Material rac-trans-4-Defluoro-4-ethoxy Paroxetine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.