missing translation for 'onlineSavingsMsg'
Learn More

Desthiazolylmethyloxycarbonyl Ritonavir-d6, TRC

Product Code. 30355052
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355052 1 mg 1mg
30356071 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30355052 Supplier Toronto Research Chemicals Supplier No. TRCD2974321MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217772-02-5
Molecular Formula C32 D6 H39 N5 O3 S
Molecular Weight (g/mol) 585.83
Synonym (2S)-N-[(1S,3S,4S)-4-Amino-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]-3-methyl-2-[[[methyl[[2-(1-methylethyl-d6)-4-thiazolyl]methyl]amino]carbonyl]amino]butanamide
IUPAC Name (2S)-N-[(1S,3S,4S)-4-amino-1-benzyl-3-hydroxy-5-phenyl-pentyl]-3-methyl-2-[[methyl-[[2-[2,2,2-trideuterio-1-(trideuteriomethyl)ethyl]thiazol-4-yl]methyl]carbamoyl]amino]butanamide
SMILES [2H]C([2H])([2H])C(c1nc(CN(C)C(=O)N[C@@H](C(C)C)C(=O)N[C@H](C[C@H](O)[C@@H](N)Cc2ccccc2)Cc3ccccc3)cs1)C([2H])([2H])[2H]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C32H45N5O3S/c1-21(2)29(36-32(40)37(5)19-26-20-41-31(35-26)22(3)4)30(39)34-25(16-23-12-8-6-9-13-23)18-28(38)27(33)17-24-14-10-7-11-15-24/h6-15,20-22,25,27-29,38H,16-19,33H2,1-5H3,(H,34,39)(H,36,40)/t25-,27-,28-,29-/m0/s1/i3D3,4D3
Formula Weight 585.362
Chemical Name or Material Desthiazolylmethyloxycarbonyl Ritonavir thiazolyl-isopropyl-1,1,1,3,3,3-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.