missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Chlorobenzyl 4-Fluorophenyl Ketone, TRC

Product Code. 30355335
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355335 250 mg 250mg
30353921 100 mg 100mg
30362117 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30355335 Supplier Toronto Research Chemicals Supplier No. TRCC428980250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62148-67-8
Molecular Formula C14H10ClFO
Molecular Weight (g/mol) 248.68
Synonym 2-Chloro-1-(4-fluorophenyl)-2-phenylethanone
IUPAC Name 2-chloro-1-(4-fluorophenyl)-2-phenylethanone
SMILES Fc1ccc(cc1)C(=O)C(Cl)c2ccccc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H10ClFO/c15-13(10-4-2-1-3-5-10)14(17)11-6-8-12(16)9-7-11/h1-9,13H
Formula Weight 248.0404
Chemical Name or Material alpha-Chlorobenzyl 4-Fluorophenyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.