missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenol-d4, TRC

Product Code. 30356555
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356555 100 mg 100mg
30360887 25 mg -
30364632 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30356555 Supplier Toronto Research Chemicals Supplier No. TRCC375042100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 285132-91-4
Molecular Formula C6 2H4 H Cl O
Molecular Weight (g/mol) 132.58
Synonym Phen-2,3,5,6-d4-ol, 4-chloro- (9CI), 4-Chlorophenol-D4, p-Chlorophenol-d4, 4-Chlorophenol D4 (phenyl D4)
IUPAC Name 4-chloro-2,3,5,6-tetradeuteriophenol
SMILES [2H]c1c([2H])c(Cl)c([2H])c([2H])c1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H5ClO/c7-5-1-3-6(8)4-2-5/h1-4,8H/i1D,2D,3D,4D
Formula Weight 132.028
Chemical Name or Material 4-Chlorophenol ring-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.