missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diaminohexane-d12 Dihydrochloride, TRC

Product Code. 30357147
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357147 5 mg 5mg
30366773 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30357147 Supplier Toronto Research Chemicals Supplier No. TRCD4161825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6H4D12N2• HCl
Molecular Weight (g/mol) 201.20046
Synonym 1,6-Diamino-n-hexane-d12, 1,6-Hexylenediamine-d12, Advancure-12, HMDA-d12, Hexamethylenediamine-d12, Hexylenediamine-d12, Hi Perm-d12, RT Advancure, HD-d12, α,ω-Hexanediamine-d12
IUPAC Name hexane-d12-1,6-diamine di-hydrochloride
SMILES [2H]C(C([2H])(C([2H])(C([2H])(N)[2H])[2H])[2H])(C([2H])(C([2H])(N)[2H])[2H])[2H].Cl.Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H16N2.2ClH/c7-5-3-1-2-4-6-8;;/h1-8H2;2*1H/i1D2,2D2,3D2,4D2,5D2,6D2;;
Formula Weight 200.17
Chemical Name or Material 1,6-Diaminohexane-d12 Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.