missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-2-hydroxybenzaldehyde, TRC

Product Code. 30358091
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358091 500 mg 500mg
30351252 50 mg -
30363005 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358091 Supplier Toronto Research Chemicals Supplier No. TRCC413400500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23731-06-8
Molecular Formula C8H7O2Cl
Molecular Weight (g/mol) 170.59
IUPAC Name 5-(chloromethyl)-2-hydroxybenzaldehyde
SMILES Oc1ccc(CCl)cc1C=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H7ClO2/c9-4-6-1-2-8(11)7(3-6)5-10/h1-3,5,11H,4H2
Formula Weight 170.0135
Chemical Name or Material 5-(Chloromethyl)-2-hydroxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.