missing translation for 'onlineSavingsMsg'
Learn More

5(6)-Carboxytetramethylrhodamine Succinimidyl Ester (Technical Grade), TRC

Product Code. 30358460
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358460 10 mg -
30359092 25 mg 25mg
30367042 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358460 Supplier Toronto Research Chemicals Supplier No. TRCC18265010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 246256-50-8
Molecular Formula C24 H22 N2 O3 . C6 H7 N O4
Molecular Weight (g/mol) 543.567
Synonym 5,6-Carboxytetramethylrhodamine Succinimidyl Ester, 5/6-TAMRA-SE, NHS-Rhodamine
IUPAC Name 2-[3,6-bis(dimethylamino)xanthen-10-ium-9-yl]benzoate;(2,5-dioxopyrrolidin-1-yl) acetate
SMILES CN(C)c1ccc2c(c3ccccc3C(=O)[O-])c4ccc(cc4[o+]c2c1)N(C)C.CC(=O)ON5C(=O)CCC5=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H22N2O3.C6H7NO4/c1-25(2)15-9-11-19-21(13-15)29-22-14-16(26(3)4)10-12-20(22)23(19)17-7-5-6-8-18(17)24(27)28;1-4(8)11-7-5(9)2-3-6(7)10/h5-14H,1-4H3;2-3H2,1H3
Formula Weight 543.201
Chemical Name or Material 5(6)-Carboxytetramethylrhodamine Succinimidyl Ester (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.