missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Dehydrocortisol 21-Hydrogen Succinate, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Dehydrocortisol 21-Hydrogen Succinate |
| Quantity | 50 mg |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H32 O8 |
| Formula Weight | 460.21 |
| Molecular Weight (g/mol) | 460.517 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?