missing translation for 'onlineSavingsMsg'
Learn More

N-Despropyl Ropinirole, TRC

Product Code. 30359624
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359624 10 mg 10mg
30368618 25 mg 25mg
30367085 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359624 Supplier Toronto Research Chemicals Supplier No. TRCD29738010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106916-16-9
Molecular Formula C13 H18 N2 O
Molecular Weight (g/mol) 218.29
Synonym 2H-Indol-2-one, 1,3-dihydro-4-[2-(propylamino)ethyl]-, 1,3-Dihydro-4-[2-(propylamino)ethyl]-2H-indol-2-one, 4-[2-(Propylamino)ethyl]-1,3-dihydro-2H-indol-2-one, SKF 104557, 4-[2-(Propylamino)ethyl]-1,3-dihydro-2H-indol-2-one
IUPAC Name 4-[2-(propylamino)ethyl]-1,3-dihydroindol-2-one
SMILES CCCNCCc1cccc2NC(=O)Cc12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H18N2O/c1-2-7-14-8-6-10-4-3-5-12-11(10)9-13(16)15-12/h3-5,14H,2,6-9H2,1H3,(H,15,16)
Formula Weight 218.1419
Chemical Name or Material N-Despropyl Ropinirole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.