missing translation for 'onlineSavingsMsg'
Learn More

Curcumin Beta-D-Glucuronide Triacetate Methyl Ester, TRC

Product Code. 30359628
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359628 2.5 mg 2.5mg
30351499 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30359628 Supplier Toronto Research Chemicals Supplier No. TRCC8385152.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1529769-81-0
Molecular Formula C34H36O15
Molecular Weight (g/mol) 684.64
Synonym 4-[(1E,6E)-7-(4-Hydroxy-3-methoxyphenyl)-3,5-dioxo-1,6-heptadien-1-yl]-2-methoxyphenyl beta-D-Glucopyranosiduronic Acid Triacetate Methyl Ester
IUPAC Name methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-[4-[(1E,6E)-7-(4-hydroxy-3-methoxyphenyl)-3,5-dioxohepta-1,6-dienyl]-2-methoxyphenoxy]oxane-2-carboxylate
SMILES COC(=O)[C@H]1O[C@@H](Oc2ccc(\C=C\C(=O)CC(=O)\C=C\c3ccc(O)c(OC)c3)cc2OC)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C34H36O15/c1-18(35)45-29-30(46-19(2)36)32(47-20(3)37)34(49-31(29)33(41)44-6)48-26-14-10-22(16-28(26)43-5)8-12-24(39)17-23(38)11-7-21-9-13-25(40)27(15-21)42-4/h7-16,29-32,34,40H,17H2,1-6H3/b11-7+,12-8+/t29-,30-,31-,32+,34+/m0/s1
Formula Weight 684.2054
Chemical Name or Material Curcumin beta-D-Glucuronide Triacetate Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.