missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroprionaldehyde Diethyl Acetal, Technical grade (Stabilized), TRC

Product Code. 30363118
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363118 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363118 Supplier Toronto Research Chemicals Supplier No. TRCC3797955ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-chloro-1,1-diethoxypropane
CAS 35573-93-4
Molecular Formula C7 H15 Cl O2
Quantity 5 mL
Synonym 1-Chloro-3,3-diethoxypropane, 3,3-Diethoxypropyl chloride, 3-Chloro-1,1-diethoxypropane, 3-Chloroprionaldehyde Diethyl Acetal, 3-Chloropropanal Diethyl Acetal, NSC 60136, β-Chloropropanal Diethyl Acetal, β-Chloropropionaldehyde Diethyl Acetal
InChI Formula InChI=1S/C7H15ClO2/c1-3-9-7(5-6-8)10-4-2/h7H,3-6H2,1-2H3
SMILES CCOC(CCCl)OCC
IUPAC Name 3-chloro-1,1-diethoxypropane
Molecular Weight (g/mol) 166.646
Formula Weight 166.076
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.