missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone 21-[O'-(2-Hydroxyethyl)]phosphate Ester, TRC

Product Code. 30363839
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.50mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363839 50 mg 50mg
30354492 10 mg 10mg
30351765 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363839 Supplier Toronto Research Chemicals Supplier No. TRCD29876050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dexamethasone 21-[O'-(2-Hydroxyethyl)]phosphate ester
Analyte or Component Names Dexamethasone 21-[O'-(2-Hydroxyethyl)]phosphate ester
Shipping Condition Room Temperature
Molecular Formula C24 H34 F O9 P
InChI Formula InChI=1S/C24H34FO9P/c1-14-10-18-17-5-4-15-11-16(27)6-7-21(15,2)23(17,25)19(28)12-22(18,3)24(14,30)20(29)13-34-35(31,32)33-9-8-26/h6-7,11,14,17-19,26,28,30H,4-5,8-10,12-13H2,1-3H3,(H,31,32)/t14-,17+,18+,19+,21+,22+,23+,24+/m1/s1
Solution Type Neat
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP(=O)(O)OCCO
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2-hydroxyethyl hydrogen phosphate
Industry Type Forensics
Molecular Weight (g/mol) 516.49
Quantity 50 mg
Formula Weight 516.1924
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.