missing translation for 'onlineSavingsMsg'
Learn More

4’-Chloro Clomiphene Citrate(E/Z Mixture), TRC

Product Code. 30367437
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367437 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367437 Supplier Toronto Research Chemicals Supplier No. TRCC36565025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Chloro Clomiphene Citrate (EZ mixture)
CAS 14158-75-9
Molecular Formula C26 H27 Cl2 N O . C6 H8 O7
Quantity 25 mg
Synonym 2-[4-[2-chloro-2-(4-chlorophenyl)-1-phenylethenyl]phenoxy]-N,N-diethylethanamine 2-hydroxy-1,2,3-propanetricarboxylate (1:1), 2-[p-(p,β-dichloro-α-phenylstyryl)phenoxy]triethylamine Citrate (1:1) (8CI), 2-[4-[2-chloro-2-(4-chlorophenyl)-1-phenylethenyl]phenoxy]-N,N-diethylethanamine Citrate, Clomifene Citrate Imp. F (EP) as Citrate
InChI Formula InChI=1S/C26H27Cl2NO.C6H8O7/c1-3-29(4-2)18-19-30-24-16-12-21(13-17-24)25(20-8-6-5-7-9-20)26(28)22-10-14-23(27)15-11-22;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-17H,3-4,18-19H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
SMILES CCN(CC)CCOc1ccc(cc1)C(=C(Cl)c2ccc(Cl)cc2)c3ccccc3.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC Name 2-[4-[2-chloro-2-(4-chlorophenyl)-1-phenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 632.53
Formula Weight 631.174
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.