missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Olmesartan Medoxomil, TRC

Product Code. 30369844
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369844 10 mg
30373679 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30369844 Supplier Toronto Research Chemicals Supplier No. TRCE92533510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1378863-74-1
Molecular Formula C30 H32 N6 O6
Molecular Weight (g/mol) 572.61
Synonym 1H-Imidazole-5-carboxylic acid, 4-(1-hydroxy-1-methylpropyl)-2-propyl-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester
IUPAC Name (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 5-(2-hydroxybutan-2-yl)-2-propyl-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylate
SMILES CCCc1nc(c(C(=O)OCC2=C(C)OC(=O)O2)n1Cc3ccc(cc3)c4ccccc4c5nnn[nH]5)C(C)(O)CC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C30H32N6O6/c1-5-9-24-31-26(30(4,39)6-2)25(28(37)40-17-23-18(3)41-29(38)42-23)36(24)16-19-12-14-20(15-13-19)21-10-7-8-11-22(21)27-32-34-35-33-27/h7-8,10-15,39H,5-6,9,16-17H2,1-4H3,(H,32,33,34,35)
Formula Weight 572.2383
Chemical Name or Material Ethyl Olmesartan Medoxomil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.