missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylhexadecylamine, TRC

Product Code. 30373421
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373421 10 g 10g
30371787 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30373421 Supplier Toronto Research Chemicals Supplier No. TRCD46183510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-69-6
Molecular Formula C18 H39 N
Molecular Weight (g/mol) 269.51
Synonym 1-Hexadecanamine, N,N-dimethyl-, Hexadecylamine, N,N-dimethyl- (6CI,7CI,8CI), N,N-Dimethyl-1-hexadecanamine, ADMA 16, AT 1695, Armeen 2M16D, Armeen DM 16D, Bairdcat B 16, Barlene 16S, Cetyldimethylamine, Dabco B 16, Dimethylcetylamine, Dimethylhexadecylamine, Dimethylpalmitylamine, Farmin DM 60, Farmin DM 6098, Genamin 16R302D, Hexadecyldimethylamine, IPL 67, Lipomin DM 16D, N,N-Dimethyl-n-hexadecylamine, N,N-Dimethylcetylamine, N,N-Dimethylhexadecylamine, N,N-Dimethylpalmitylamine, N-Hexadecyldimethylamine, NSC 404177, NSC 8493, Nissan Tertiary Amine PB, Palmityldimethylamine
IUPAC Name N,N-dimethylhexadecan-1-amine
SMILES CCCCCCCCCCCCCCCCN(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C18H39N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(2)3/h4-18H2,1-3H3
Formula Weight 269.3083
Chemical Name or Material N,N-Dimethylhexadecan-1-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.