missing translation for 'onlineSavingsMsg'
Learn More

Dopamine-d3 Hydrochloride, TRC

Product Code. 30374576
Change view
Click to view available options
Quantity:
1.5 mg
3 mg
15 mg
Unit Size:
1.5mg
3mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374576 3 mg 3mg
30377133 1.5 mg 1.5mg
30371785 15 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30374576 Supplier Toronto Research Chemicals Supplier No. TRCD5337853MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53587-30-7
Molecular Formula C8 D3 H8 N O2 . Cl H
Molecular Weight (g/mol) 192.66
Synonym 2-(3,4-Dihydroxyphenyl-d3)ethylamine HCl
IUPAC Name 4-(2-aminoethyl)-3,5,6-trideuteriobenzene-1,2-diol;hydrochloride
SMILES Cl.[2H]c1c([2H])c(CCN)c([2H])c(O)c1O

Specifications

Quantity 3 mg
InChI Formula InChI=1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H/i1D,2D,5D;
Formula Weight 192.0745
Percent Purity >95%
Chemical Name or Material Dopamine-d3 Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.