missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ditridecyl Phthalate, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(tridecyl) phthalate |
| CAS | 119-06-2 |
| Molecular Formula | C34 H58 O4 |
| Quantity | 25 mg |
| Synonym | 1,2-Benzenedicarboxylic acid, ditridecyl ester (9CI), Phthalic acid, ditridecyl ester (6CI,7CI,8CI), 1-Tridecanol, phthalate (2:1) (8CI), Bis(tridecyl) phthalate, Di-n-tridecyl phthalate, Ditridecyl phthalate, Hatcol 2922, Hexaplas DTDP, Jayflex DTDP, Polycizer 962BPA, Staflex DTDP, Truflex DTDP |
| InChI Formula | InChI=1S/C34H58O4/c1-3-5-7-9-11-13-15-17-19-21-25-29-37-33(35)31-27-23-24-28-32(31)34(36)38-30-26-22-20-18-16-14-12-10-8-6-4-2/h23-24,27-28H,3-22,25-26,29-30H2,1-2H3 |
| SMILES | CCCCCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCCCCCC |
| IUPAC Name | ditridecyl benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 530.82 |
| Formula Weight | 530.4335 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?