missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dipalmitoyl-2-oleoyl Glycerol, TRC

Product Code. 30380218
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380218 100 mg -
30378143 25 mg -
30382015 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380218 Supplier Toronto Research Chemicals Supplier No. TRCD486845100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2190-25-2
Molecular Formula C53H100O6
Molecular Weight (g/mol) 833.36
Synonym (9Z)-9-Octadecenoic Acid 2-[(1-Oxohexadecyl)oxy]-1-[[(1-oxohexadecyl)oxy]methyl]ethyl Ester, 1,3-Dipalmito-2-olein, 1,3-Dipalmitoyl-2-olein, 1,3-Dipalmitoyl-2-oleoylglycerin, 1,3-Dipalmitoyl-2-oleoylglycerol, 1-Palmito-2-oleo-3-palmitin, 2-Oleo-1,3-dipalmitin, 2-Oleodipalmitin, 2-Oleoyl-1,3-dipalmitin, 2-Oleoyl-1,3-dipalmitoylglycerin, 2-Oleoyl-1,3-dipalmitoylglycerol, Glycerin-1-palmitate-2-oleate-3-palmitate, Glycerol 1,3-Dipalmitate 2-oleate, Glyceryl 2-oleate-1,3-dipalmitate, POP
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCC\C=C\CCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C53H100O6/c1-4-7-10-13-16-19-22-25-26-29-32-35-38-41-44-47-53(56)59-50(48-57-51(54)45-42-39-36-33-30-27-23-20-17-14-11-8-5-2)49-58-52(55)46-43-40-37-34-31-28-24-21-18-15-12-9-6-3/h25-26,50H,4-24,27-49H2,1-3H3/b26-25-
Formula Weight 832.752
Chemical Name or Material 1,3-Dipalmitoyl-2-oleoyl Glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.