missing translation for 'onlineSavingsMsg'
Learn More
Learn More
15-Keto Prostaglandin E1, TRC
Specifications
Specifications
| Chemical Name or Material | 15-Ketoprostaglandin E1 |
| CAS | 22973-19-9 |
| Molecular Formula | C20 H32 O5 |
| Quantity | 5 mg |
| Synonym | Prost-13-en-1-oic acid, 11-hydroxy-9,15-dioxo-, (11α,13E)-, Cyclopentaneheptanoic acid, 3-hydroxy-5-oxo-2-(3-oxo-1-octenyl)- (7CI), Cyclopentaneheptanoic acid, 3-hydroxy-5-oxo-2-(3-oxo-1-octenyl)-, stereoisomer (8CI), (11α,13E)-11-Hydroxy-9,15-dioxoprost-13-en-1-oic acid, 15-Keto-PGE1, 15-Ketoprostaglandin E1, 15-Oxo-PGE1, 15-Oxoprostaglandin E1, 15-keto-PGE1, 15-keto-Prostaglandin E1, U 22409, 7-[(1R,2R,3R)-3-Hydroxy-2-[(1E)-3-oxooct-1-enyl]-5-oxocyclopentyl]heptanoic Acid |
| InChI Formula | InChI=1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,16-17,19,23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t16-,17-,19-/m1/s1 |
| SMILES | CCCCCC(=O)\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(=O)O |
| IUPAC Name | 7-[(1R,2R,3R)-3-hydroxy-5-oxo-2-[(E)-3-oxooct-1-enyl]cyclopentyl]heptanoic acid |
| Molecular Weight (g/mol) | 352.47 |
| Formula Weight | 352.225 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?