missing translation for 'onlineSavingsMsg'
Learn More

2,3-O-Isopropylidene-3-hydroxymethyl-D-ribose, TRC

Product Code. 30396445
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396445 2.5 g 2.5g
30404094 250 mg 250mg
30399447 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396445 Supplier Toronto Research Chemicals Supplier No. TRCI8254602.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-O-Isopropylidene-3-hydroxymethyl-D-ribose
Analyte or Component Names 2,3-O-Isopropylidene-3-hydroxymethyl-D-ribose
Shipping Condition Room Temperature
Molecular Formula C29 H50 O5
Synonym (4-Hydroxy-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxole-3a,6-diyl)dimethanol
InChI Formula InChI=1S/C29H50O5/c1-7-17(15(2)3)27(34)26(33)16(4)19-8-9-20-18-12-23(30)22-13-24(31)25(32)14-29(22,6)21(18)10-11-28(19,20)5/h15-22,24-27,31-34H,7-14H2,1-6H3/t16-,17+,18+,19+,20-,21-,22+,24-,25+,26-,27-,28-,29-/m1/s1
Solution Type Neat
SMILES CC[C@@H](C(C)C)[C@@H](O)[C@H](O)[C@H](C)[C@@H]1CC[C@@H]2[C@@H]3CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@@H]3CC[C@]12C
IUPAC Name (2S,3R,5R,8S,9R,10R,13S,14R,17S)-17-[(2R,3R,4R,5S)-5-ethyl-3,4-dihydroxy-6-methylheptan-2-yl]-2,3-dihydroxy-10,13-dimethyl-1,2,3,4,5,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-6-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 478.704
Quantity 2.5 g
Formula Weight 478.366
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.