missing translation for 'onlineSavingsMsg'
Learn More

Hygromycin B Deuterated (d4 major), TRC

Product Code. 30399512
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.5mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399512 2.5 mg 2.5mg
30392490 10 mg 10mg
30403671 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30399512 Supplier Toronto Research Chemicals Supplier No. TRCH9976042.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hygromycin B D4
Molecular Formula C20H33D4N3O13
Quantity 2.5 mg
Synonym O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene-(1→2-3)-O-β-D-talopyranosyl-(1→5)-2-deoxy-N3-methyl-D-streptamine Deuterated, Antihelmycin Deuterated, Hygromix 2.4 Deuterated, Hygromix-8 Deuterated, Hygrovetin Deuterated, Hygrovetine Deuterated
InChI Formula InChI=1S/C20H37N3O13/c1-23-7-2-5(21)9(26)15(10(7)27)33-19-17-16(11(28)8(4-25)32-19)35-20(36-17)18(31)13(30)12(29)14(34-20)6(22)3-24/h5-19,23-31H,2-4,21-22H2,1H3/t5-,6?,7-,8+,9-,10+,11+,12+,13+,14-,15-,16+,17+,18-,19?,20-/m0/s1
SMILES O[C@H]1[C@@H](OC2O[C@H](CO)[C@@H](O)[C@@H]3[C@H]2O[C@@]4(O[C@@H](C(N)CO)[C@H](O)[C@@H](O)[C@@H]4O)O3)[C@@H](O)[C@@H](N)C[C@@H]1NC
Molecular Weight (g/mol) 531.54
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.