missing translation for 'onlineSavingsMsg'
Learn More

[3-Fluoro-4-(4-morpholinyl)phenyl]carbamic Acid Phenylmethyl Ester, TRC

Product Code. 30399960
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399960 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399960 Supplier Toronto Research Chemicals Supplier No. TRCF5949801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl 3-Fluoro-4-(4-morpholinyl)phenylcarbamate
CAS 168828-81-7
Molecular Formula C18 H19 F N2 O3
Quantity 1 g
Synonym N-[3-Fluoro-4-(4-morpholinyl)phenyl]carbamic Acid Phenylmethyl Ester, Carbamic acid, [3-fluoro-4-(4-morpholinyl)phenyl]-, phenylmethyl ester (9CI), (3-Fluoro-4-morpholin-4-ylphenyl)carbamic acid benzyl ester, Benzyl 3-fluoro-4-(4-morpholinyl)phenylcarbamate, N-(Benzyloxycarbonyl)-3-fluoro-4-(4-morpholinyl)aniline, N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline, [3-Fluoro-4-(4-morpholinyl)phenyl]carbamic acid phenylmethyl ester
InChI Formula InChI=1S/C18H19FN2O3/c19-16-12-15(6-7-17(16)21-8-10-23-11-9-21)20-18(22)24-13-14-4-2-1-3-5-14/h1-7,12H,8-11,13H2,(H,20,22)
SMILES Fc1cc(NC(=O)OCc2ccccc2)ccc1N3CCOCC3
IUPAC Name benzyl N-(3-fluoro-4-morpholin-4-ylphenyl)carbamate
Molecular Weight (g/mol) 330.35
Formula Weight 330.138
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.