missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Isopropylbenzoic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Isopropylbenzoic acid |
| CAS | 5651-47-8 |
| Molecular Formula | C10 H12 O2 |
| Quantity | 100 mg |
| Synonym | Benzoic acid, m-isopropyl- (6CI,7CI,8CI), 3-(1-Methylethyl)benzoic acid, 3-(Propan-2-yl)benzoic acid, 3-Propane-2-ylbenzoic acid, NSC 59899, m-Cumic acid, m-Isopropylbenzoic acid |
| InChI Formula | InChI=1S/C10H12O2/c1-7(2)8-4-3-5-9(6-8)10(11)12/h3-7H,1-2H3,(H,11,12) |
| SMILES | CC(C)c1cccc(c1)C(=O)O |
| IUPAC Name | 3-propan-2-ylbenzoic acid |
| Molecular Weight (g/mol) | 164.201 |
| Formula Weight | 164.084 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?