missing translation for 'onlineSavingsMsg'
Learn More

Isoanastrozole Diamide, TRC

Product Code. 30400838
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400838 5 mg 5mg
30405476 2.5 mg 2.5mg
30400569 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400838 Supplier Toronto Research Chemicals Supplier No. TRCI7799755MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoanastrozole Diamide
Analyte or Component Names Isoanastrozole Diamide
Shipping Condition Room Temperature
Molecular Formula C17 H23 N5 O2
InChI Formula InChI=1S/C17H23N5O2/c1-16(2,14(18)23)12-5-11(8-22-9-20-21-10-22)6-13(7-12)17(3,4)15(19)24/h5-7,9-10H,8H2,1-4H3,(H2,18,23)(H2,19,24)
Solution Type Neat
SMILES CC(C)(C(=O)N)c1cc(Cn2cnnc2)cc(c1)C(C)(C)C(=O)N
IUPAC Name 2-[3-(1-amino-2-methyl-1-oxopropan-2-yl)-5-(1,2,4-triazol-4-ylmethyl)phenyl]-2-methylpropanamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 329.397
Quantity 5 mg
Formula Weight 329.185
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.