missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenylboronic acid, TRC

Product Code. 30401922
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401922 500 mg -
30405908 250 mg 250mg
30389267 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401922 Supplier Toronto Research Chemicals Supplier No. TRCH952913500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89466-08-0
Molecular Formula C6H7BO3
Molecular Weight (g/mol) 137.93
IUPAC Name (2-hydroxyphenyl)boronic acid
SMILES OB(O)c1ccccc1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H7BO3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8-10H
Formula Weight 138.0488
Chemical Name or Material 2-Hydroxyphenylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.