missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-6-nitroanisole, TRC

Product Code. 30401958
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401958 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401958 Supplier Toronto Research Chemicals Supplier No. TRCF59377010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Fluoro-6-nitroanisole
CAS 448-19-1
Molecular Formula C7H6FNO3
Quantity 10 g
Synonym 1-Fluoro-3-methoxy-4-nitrobenzene, 2-Methoxy-4-fluoronitrobenzene, 2-Nitro-5-fluoroanisole, 3-Methoxy-4-nitrofluorobenzene, 4-Fluoro-2-(methyloxy)-1-nitrobenzene, 4-Fluoro-2-methoxy-1-nitrobenzene, 4-Fluoro-2-methoxynitrobenzene, 5-Fluoro-2-nitroanisole, 5-Fluoro-2-nitrophenyl Methyl Ether
InChI Formula InChI=1S/C7H6FNO3/c1-12-7-4-5(8)2-3-6(7)9(10)11/h2-4H,1H3
SMILES COc1cc(F)ccc1[N+](=O)[O-]
Molecular Weight (g/mol) 171.13
Formula Weight 171.0332
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.