missing translation for 'onlineSavingsMsg'
Learn More

(1-Propionylpiperidin-4-yl)methyl methanesulfonate, TRC

Product Code. 30469890
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469890 500 mg 500mg
30469016 50 mg 50mg
30470900 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30469890 Supplier Toronto Research Chemicals Supplier No. TRCM287645500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1357353-59-3
Molecular Formula C10H19NO4S
Molecular Weight (g/mol) 249.33
Synonym 1-[4-[[(Methylsulfonyl)oxy]methyl]-1-piperidinyl]-1-propanone
IUPAC Name (1-propanoylpiperidin-4-yl)methyl methanesulfonate
SMILES CCC(=O)N1CCC(COS(=O)(=O)C)CC1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H19NO4S/c1-3-10(12)11-6-4-9(5-7-11)8-15-16(2,13)14/h9H,3-8H2,1-2H3
Formula Weight 249.1035
Chemical Name or Material (1-Propionylpiperidin-4-yl)methyl methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.