missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Nicotinamide Adenine Dinucleotide Sodium Salt Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | beta-Nicotinamide adenine dinucleotide sodium salt hydrate |
| Molecular Formula | C21 H26 N7 O14 P2 . Na . x H2 O |
| Quantity | 100 mg |
| Synonym | Codehydrase I sodium salt hydrate, Coenzyme I sodium salt hydrate |
| InChI Formula | InChI=1S/C21H27N7O14P2.Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37);/q;+1/p-1/t10-,11-,13-,14-,15-,16-,20-,21-;/m1./s1 |
| SMILES | [Na+].NC(=O)c1ccc[n+](c1)[C@@H]2O[C@H](COP(=O)([O-])OP(=O)([O-])OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O |
| IUPAC Name | sodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methoxy-oxido-phosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methyl phosphate |
| Molecular Weight (g/mol) | 685.41 |
| Formula Weight | 685.0911 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?