missing translation for 'onlineSavingsMsg'
Learn More

Nifenazone, TRC

Product Code. 30480614
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480614 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480614 Supplier Toronto Research Chemicals Supplier No. TRCN4573005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nifenazone
CAS 2139-47-1
Molecular Formula C17 H16 N4 O2
Quantity 5 g
Synonym 3-Pyridinecarboxamide','N-(2,3-dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)-, Nicotinamide','N-antipyrinyl- (6CI,7CI,8CI), 4-(Nicotinoylamino)antipyrine, 4-Nicotinamido-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Dolongan, N-Antipyrinylnicotinamide, N-Nicotinoylaminoantipyrine, Niapyrin, Nicazane, Nicodon, Nicodone, Nicofesone, Nicophesone, Nicophezon, Nicopyron, Nicoreumal, Nicoron, Nicotazone, Nicotinic acid 4-antipyrilamide, Nicotinoylantipyrine, Nicozone, Nifenazon, Nifenazone, Nikodon, Nikofezon, Niprazine, Phenicazone, Reupiron, Thylin
InChI Formula InChI=1S/C17H16N4O2/c1-12-15(19-16(22)13-7-6-10-18-11-13)17(23)21(20(12)2)14-8-4-3-5-9-14/h3-11H,1-2H3,(H,19,22)
SMILES CN1N(C(=O)C(=C1C)NC(=O)c2cccnc2)c3ccccc3
IUPAC Name N-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)pyridine-3-carboxamide
Molecular Weight (g/mol) 308.33
Formula Weight 308.1273
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.