missing translation for 'onlineSavingsMsg'
Learn More

(+)-Noradrenaline Bitartrate, TRC

Product Code. 30481342
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481342 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481342 Supplier Toronto Research Chemicals Supplier No. TRCN661025100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-(+)-Noradrenaline Tartrate
CAS 636-88-4
Purity Grade Notes HPLC
Molecular Formula C8 H11 N O3 . C4 H6 O6
Quantity 100 mg
Synonym 1,2-Benzenediol','4-[(1S)-2-amino-1-hydroxyethyl]-','(2R,3R)-2,3-dihydroxybutanedioate (1:1), 1,2-Benzenediol','4-(2-amino-1-hydroxyethyl)-','(S)-','[R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), 1,2-Benzenediol','4-[(1S)-2-amino-1-hydroxyethyl]-','(2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt), Benzyl alcohol','α-(aminomethyl)-3,4-dihydroxy-','(+)-','tartrate (1:1) (salt), (+)-(S)-Norepinephrine bitartrate, (+)-Noradrenaline (+)-bitartrate, (+)-Noradrenaline bitartrate, (+)-Norepinephrine (+)-bitartrate, (+)-Norepinephrine bitartrate, D-Norepinephrine bitartrate, D-Norepinephrine d-bitartrate, S-(+)-Norepinephrine bitartrate, d-Noradrenaline bitartrate, d-Noradrenaline hydrogen tartrate
InChI Formula InChI=1S/C8H11NO3.C4H6O6/c9-4-8(12)5-1-2-6(10)7(11)3-5;5-1(3(7)8)2(6)4(9)10/h1-3,8,10-12H,4,9H2;1-2,5-6H,(H,7,8)(H,9,10)/t8-;1-,2-/m11/s1
SMILES NC[C@@H](O)c1ccc(O)c(O)c1.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC Name 4-[(1S)-2-amino-1-hydroxyethyl]benzene-1,2-diol;(2R,3R)-2,3-dihydroxybutanedioic acid
Molecular Weight (g/mol) 319.26
Formula Weight 319.0903
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.